Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMR1NB
Entrez Gene ID
613334
Gene Name
FMR1 neighbor
Gene Aliases
-
Chromosome Location
Chr.X: 30683591 - 30723111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102262.2 NP_001095732.1 1 - 2 383 87
GenBank mRNA BC149653.1 - 1 - 2 384 87

View Full Product Details