Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP7
Entrez Gene ID
613606
Gene Name
A-kinase anchoring protein 7
Gene Aliases
-
Chromosome Location
Chr.9: 69202949 - 69325809 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102266.1 NP_001095736.1 2 - 3 176 98
GenBank mRNA BC148038.1 - 2 - 3 176 98

View Full Product Details