Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC47
Entrez Gene ID
616209
Gene Name
leucine rich repeat containing 47
Gene Aliases
-
Chromosome Location
Chr.16: 49273571 - 49284794 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102298.1 NP_001095768.1 6 - 7 1500 108
GenBank mRNA BC150059.1 - 6 - 7 1500 108

View Full Product Details