Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPRA1
Entrez Gene ID
617772
Gene Name
transmembrane protein adipocyte associated 1
Gene Aliases
GPR175
Chromosome Location
Chr.22: 59921386 - 59929765 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102314.1 NP_001095784.1 4 - 5 588 95
GenBank mRNA BC149762.1 - 4 - 5 588 95

View Full Product Details