Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR32
Entrez Gene ID
783116
Gene Name
proline rich 32
Gene Aliases
CXHXorf64
Chromosome Location
Chr.X: 11131753 - 11133670 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099188.1 NP_001092658.1 1 - 2 107 68
GenBank mRNA BC142150.1 - 1 - 2 107 68

View Full Product Details