Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL16
Entrez Gene ID
789464
Gene Name
F-box and leucine rich repeat protein 16
Gene Aliases
-
Chromosome Location
Chr.25: 595081 - 606604 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099212.1 NP_001092682.1 4 - 5 1540 89
GenBank mRNA BC142518.1 - 4 - 5 1540 89

View Full Product Details