Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR5
Entrez Gene ID
788925
Gene Name
proline rich 5
Gene Aliases
-
Chromosome Location
Chr.5: 115267982 - 115320964 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101305.1 NP_001094775.1 4 - 5 297 58
GenBank mRNA BC146088.1 - 4 - 5 297 58

View Full Product Details