Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM69
Entrez Gene ID
785268
Gene Name
transmembrane protein 69
Gene Aliases
-
Chromosome Location
Chr.3: 100376386 - 100380650 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102338.1 NP_001095808.1 2 - 3 196 67
GenBank mRNA BC149737.1 - 2 - 3 196 67

View Full Product Details