The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102351.1 | NP_001095821.1 | 2 - 3 | 338 | 71 |
GenBank mRNA | BC148938.1 | - | 2 - 3 | 338 | 71 |