Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H16orf70
Entrez Gene ID
100125939
Gene Name
chromosome 18 C16orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 34758653 - 34788130 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105502.1 NP_001098972.1 10 - 11 902 75
GenBank mRNA BC142370.1 - 10 - 11 902 75

View Full Product Details