Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRL1
Entrez Gene ID
788252
Gene Name
adhesion G protein-coupled receptor L1
Gene Aliases
LPHN1
Chromosome Location
Chr.7: 11510479 - 11560787 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113728.1 NP_001107200.1 23 - 24 4124 152
GenBank mRNA AF111097.1 - 22 - 23 4109 152
AF111098.1 - 23 - 24 4124 152

View Full Product Details