Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ST6GAL2
Entrez Gene ID
493992
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Gene Aliases
SIAT2, ST6Gal II, ST6GalII
Chromosome Location
Chr.11: 45551844 - 45610884 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008668.2 NP_001008668.2 6 - 6 2452 155
GenBank mRNA BC140674.1 - 6 - 6 2452 155

View Full Product Details