Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCIN
Entrez Gene ID
281041
Gene Name
calicin
Gene Aliases
-
Chromosome Location
Chr.8: 60444357 - 60448254 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174005.4 NP_776430.2 1 - 1 1652 151
GenBank mRNA BC111363.1 - 1 - 1 1653 151
Z46968.1 - - 1605 151

View Full Product Details