Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO10
Entrez Gene ID
781854
Gene Name
F-box protein 10
Gene Aliases
-
Chromosome Location
Chr.8: 61633981 - 61676635 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144076.1 NP_001137548.1 4 - 5 1696 60

View Full Product Details