Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANX3
Entrez Gene ID
538977
Gene Name
pannexin 3
Gene Aliases
-
Chromosome Location
Chr.29: 28089645 - 28097175 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144084.2 NP_001137556.1 2 - 3 329 62

View Full Product Details