Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXW12
Entrez Gene ID
528919
Gene Name
F-box and WD repeat domain containing 12
Gene Aliases
-
Chromosome Location
Chr.8: 90070232 - 90104626 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128501.2 NP_001121973.1 6 - 7 798 62
GenBank mRNA BC103020.1 - 6 - 7 799 62

View Full Product Details