Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDAH1
Entrez Gene ID
537391
Gene Name
dimethylarginine dimethylaminohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.3: 58618306 - 58775122 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102201.2 NP_001095671.1 3 - 4 582 81
GenBank mRNA BC107532.1 - 3 - 4 581 81
BC149998.1 - 3 - 4 583 81

View Full Product Details