Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SELENOV
Entrez Gene ID
615288
Gene Name
selenoprotein V
Gene Aliases
SELV
Chromosome Location
Chr.18: 49184744 - 49190099 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163244.2 NP_001156716.1 2 - 3 1008 64
GenBank mRNA DT829759.1 - 3 - 4 296 64
HX928062.1 - 2 - 3 277 64

View Full Product Details