The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163244.2 | NP_001156716.1 | 3 - 4 | 1056 | 68 |
GenBank mRNA | DT829759.1 | - | 4 - 5 | 344 | 68 |
HX928062.1 | - | 3 - 4 | 325 | 68 |