Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
513688
Gene Name
kizuna centrosomal protein
Gene Aliases
C13H20orf19, NCRNA00153, PLK1S1
Chromosome Location
Chr.13: 40490248 - 40583450 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163458.1 NP_001156930.1 4 - 5 460 107
GenBank mRNA BC126681.1 - 4 - 5 400 107

View Full Product Details