Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C1A
Entrez Gene ID
504454
Gene Name
5'-nucleotidase, cytosolic IA
Gene Aliases
-
Chromosome Location
Chr.3: 106403642 - 106419979 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205546.3 NP_001192475.1 1 - 2 199 53

View Full Product Details