Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A28
Entrez Gene ID
538529
Gene Name
solute carrier family 25 member 28
Gene Aliases
-
Chromosome Location
Chr.26: 20596417 - 20607094 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205552.1 NP_001192481.1 1 - 2 387 62

View Full Product Details