Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND6
Entrez Gene ID
504789
Gene Name
BEN domain containing 6
Gene Aliases
-
Chromosome Location
Chr.23: 3189359 - 3252943 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205554.1 NP_001192483.1 5 - 6 826 79

View Full Product Details