Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSPT2
Entrez Gene ID
538724
Gene Name
G1 to S phase transition 2
Gene Aliases
-
Chromosome Location
Chr.X: 89276444 - 89278960 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205578.1 NP_001192507.1 1 - 2 81 56

View Full Product Details