Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK2A
Entrez Gene ID
509304
Gene Name
divergent protein kinase domain 2A
Gene Aliases
C1H3orf58
Chromosome Location
Chr.1: 124866092 - 124888751 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205651.1 NP_001192580.1 1 - 2 1345 66

View Full Product Details