Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH2D3A
Entrez Gene ID
510173
Gene Name
SH2 domain containing 3A
Gene Aliases
-
Chromosome Location
Chr.7: 17732344 - 17747013 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205669.1 NP_001192598.1 7 - 8 1467 65
GenBank mRNA BT025405.1 - 7 - 8 1466 65

View Full Product Details