Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RWDD2A
Entrez Gene ID
539733
Gene Name
RWD domain containing 2A
Gene Aliases
-
Chromosome Location
Chr.9: 23115918 - 23121547 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205704.1 NP_001192633.1 1 - 2 75 65

View Full Product Details