Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHS
Entrez Gene ID
539895
Gene Name
NHS actin remodeling regulator
Gene Aliases
-
Chromosome Location
Chr.X: 125750192 - 126095418 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205728.1 NP_001192657.1 2 - 3 703 102

View Full Product Details