Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO33
Entrez Gene ID
539998
Gene Name
F-box protein 33
Gene Aliases
-
Chromosome Location
Chr.21: 49289714 - 49310143 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205749.1 NP_001192678.1 2 - 3 884 86

View Full Product Details