Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDNF
Entrez Gene ID
513798
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
ARMETL1
Chromosome Location
Chr.13: 29470661 - 29492325 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205769.2 NP_001192698.1 2 - 3 321 72

View Full Product Details