Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF8
Entrez Gene ID
514609
Gene Name
SLAM family member 8
Gene Aliases
-
Chromosome Location
Chr.3: 9850404 - 9860647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205794.1 NP_001192723.1 2 - 3 435 101

View Full Product Details