Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A47
Entrez Gene ID
516483
Gene Name
solute carrier family 25 member 47
Gene Aliases
-
Chromosome Location
Chr.21: 65228286 - 65233524 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205829.2 NP_001192758.2 2 - 3 70 76

View Full Product Details