Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMND5A
Entrez Gene ID
516731
Gene Name
required for meiotic nuclear division 5 homolog A
Gene Aliases
-
Chromosome Location
Chr.11: 48135799 - 48196518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205832.1 NP_001192761.1 6 - 7 852 105

View Full Product Details