Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XKR7
Entrez Gene ID
518090
Gene Name
XK related 7
Gene Aliases
-
Chromosome Location
Chr.13: 61487256 - 61511791 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205845.3 NP_001192774.1 2 - 3 789 48

View Full Product Details