Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLIS2
Entrez Gene ID
527935
Gene Name
GLIS family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.25: 3483961 - 3504210 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205948.1 NP_001192877.1 2 - 3 351 102

View Full Product Details