Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRF
Entrez Gene ID
533039
Gene Name
serum response factor
Gene Aliases
-
Chromosome Location
Chr.23: 16775130 - 16782650 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206016.1 NP_001192945.1 5 - 6 1357 86

View Full Product Details