Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM47
Entrez Gene ID
540831
Gene Name
RNA binding motif protein 47
Gene Aliases
-
Chromosome Location
Chr.6: 59387155 - 59663515 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206034.1 NP_001192963.1 5 - 6 507 116

View Full Product Details