Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIPARP
Entrez Gene ID
540975
Gene Name
TCDD inducible poly(ADP-ribose) polymerase
Gene Aliases
-
Chromosome Location
Chr.1: 110928197 - 110955891 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206048.1 NP_001192977.1 5 - 6 1743 83

View Full Product Details