Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCOR2
Entrez Gene ID
541233
Gene Name
REST corepressor 2
Gene Aliases
-
Chromosome Location
Chr.29: 42253812 - 42258568 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206075.1 NP_001193004.1 2 - 3 189 83

View Full Product Details