Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB5B
Entrez Gene ID
539150
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.5: 57321742 - 57337445 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206191.2 NP_001193120.1 6 - 7 724 119

View Full Product Details