Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO1
Entrez Gene ID
508781
Gene Name
flavin containing dimethylaniline monoxygenase 1
Gene Aliases
-
Chromosome Location
Chr.16: 38801860 - 38849076 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206199.1 NP_001193128.1 1 - 2 104 100

View Full Product Details