Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLIF
Entrez Gene ID
616252
Gene Name
cobalamin binding intrinsic factor
Gene Aliases
GIF
Chromosome Location
Chr.15: 83029815 - 83048887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206239.1 NP_001193168.1 2 - 3 295 70

View Full Product Details