Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM219B
Entrez Gene ID
617177
Gene Name
family with sequence similarity 219 member B
Gene Aliases
C21H15orf17
Chromosome Location
Chr.21: 33790038 - 33796023 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206267.1 NP_001193196.1 2 - 3 283 57

View Full Product Details