Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NYNRIN
Entrez Gene ID
516993
Gene Name
NYN domain and retroviral integrase containing
Gene Aliases
-
Chromosome Location
Chr.10: 20755336 - 20776156 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192459.2 NP_001179388.1 7 - 8 2842 77

View Full Product Details