Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC38A8
Entrez Gene ID
518625
Gene Name
solute carrier family 38 member 8
Gene Aliases
-
Chromosome Location
Chr.18: 10341525 - 10385244 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192494.1 NP_001179423.1 5 - 6 693 70

View Full Product Details