Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGV
Entrez Gene ID
519213
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Gene Aliases
-
Chromosome Location
Chr.2: 126392492 - 126402287 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192505.1 NP_001179434.1 3 - 4 262 59

View Full Product Details