Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRKB
Entrez Gene ID
519224
Gene Name
TP53RK binding protein
Gene Aliases
-
Chromosome Location
Chr.11: 10874012 - 10885257 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192507.1 NP_001179436.1 2 - 3 434 109
GenBank mRNA BC102574.1 - 2 - 3 194 109

View Full Product Details