Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL11B
Entrez Gene ID
521303
Gene Name
methyltransferase like 11B
Gene Aliases
-
Chromosome Location
Chr.16: 37760115 - 37779595 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192536.1 NP_001179465.1 1 - 2 157 91

View Full Product Details