Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
522297
Gene Name
epididymal sperm binding protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 54825102 - 54848797 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192558.1 NP_001179487.1 3 - 4 352 69

View Full Product Details