Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTN1
Entrez Gene ID
522767
Gene Name
netrin 1
Gene Aliases
-
Chromosome Location
Chr.19: 28447285 - 28652556 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192566.1 NP_001179495.1 2 - 3 1211 94

View Full Product Details